missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Scott™ PERFORMANCE Standard Toilet Tissue Rolls
Provides the comfort of home. Kimberly-Clark™ Scott™ PERFORMANCE Standard Roll Toilet Tissue dissolves in cold water and won't clog drains.
Brand: Kimberly-Clark™ 8519
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- White, perforated, 2-ply toilet tissue
- Soft and absorbent
- Dissolves in cold water so that the sewer pipes will not clog up
- Perforation allows economical consumption, suitable for cost savings and reducing the amount of waste
- ISO 14001, ISO 9001, EU Label and FSC certifie
- Ideal for washrooms in office buildings and hotels that value economy
Specifications
Toilet Tissue Roll | |
9.5 cm | |
White | |
8 x 8 Rolls x 350 Sheets |
12 cm | |
ISO 14001, ISO 9001, EU Ecolabel and FSC certified | |
12 x 9.5 cm | |
2-ply |