missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SCOTfluor lactic acid probe 510

Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
SCOTfluor lactic acid probe 510 is a red fluorescent probe for imaging lactic acid metabolism in vivo. Excitation and emission maxima (λ) are 485 and 605 nm, respectively. Suitable for use with flow cytometry and super-resolution microscopy.
Specifications
Specifications
| Chemical Name or Material | (2 S)-2-Hydroxy-3-[(7-nitro-2,1,3-benzoselenadiazol-4-yl)amino]propanoic acid |
| CAS | 2351933-31-6 |
| Quantity | 2 mg |
| Target | Fluorescent Probes |
| Molecular Formula | C9H8N4O5Se |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction