missing translation for 'onlineSavingsMsg'
Learn More

3M™ Scotch-Brite™ Diamond Floor Brush

Product Code. 17813256
Click to view available options
Grit:
1000
3000
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 17813256

Brand: 3M™ 7100262369

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Scotch-Brite™ Diamond Floor Brush is designed to clean and polish uncoated concrete, terrazzo and potentially other stone floors in one step.

  • Scotch-Brite™ Diamond Floor Brush is designed to clean and polish uncoated concrete, terrazzo and potentially other stone floors in one step.
  • It is conformable, allowing the flexible bristles to reach low spots on the floor while minimizing debris clogging.
  • No assembly or pre-soaking is required and it is easy to install on standard, low-speed equipment.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Flexible Brush
Bristle Material Bristle
Bristle Type Flexible
Color-coded No
Product Line Scotch-Brite
Tip Type Without Tip
Scent Unscented
Sterility Non-sterile
Color Gray
For Use With (Equipment) Low-Speed Machine
Grit 1000
Size 432 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.