missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Scorpio™ 19-024 Series Black Neoprene Immersion Gloves

Product Code. p-8477002 Shop All Ansell Products
Click to view available options
Size:
8
10
Unit Size:
1 pair

Product Code. 11741734

Brand: Ansell™ 19024080

Only null left
Add to basket
Add to basket

Combine the flexibility and chemical resistance of neoprene with the comfort and heat resistance of a soft thermal liner. Provides protection from intermittent contact with hot surfaces up to 180°C.

  • Chemical resistant gloves
  • Combines the flexibility and chemical resistance of neoprene with the comfort and heat resistance of a soft thermal liner
  • Designed to protect the users in applications where thermal and chemical risks are present
  • Good mechanical and chemical properties
  • Unaffected by detergents and cleaning solutions
  • Offers protection against greases and oils
  • Superior flexibility and grip

Applications:

  • Charging and blending raw materials
  • Handling materials with sharp or rough edges
  • Press work
  • Production line support and maintenance
  • Unexpected leakages spills or other releases
TRUSTED_SUSTAINABILITY

Specifications

Back Material Neoprene
Cuff Material Neoprene
Applications Industrial
Color Black
Latex Free Yes
Material Neoprene
Product Type Immersion Gloves
Size 8
Temperature (Metric) Max 180°C
Glove Cuff Color Black
Glove Cuff Style Gauntlet
Quantity 1 Pair
Resistant To Chemical resistance
Series 19-024
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.