missing translation for 'onlineSavingsMsg'
Learn More

Scopoline Methobromide, TRC

Product Code. 17914625
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17914625 5 mg 5mg
17901595 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17914625 Supplier Toronto Research Chemicals Supplier No. TRCS1999955MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1044148-31-3
Molecular Formula C9 H16 N O2 . Br
Molecular Weight (g/mol) 250.13
Synonym (2S,3aR,5S,6S,6aS)-Hexahydro-6-hydroxy-4,4-dimethyl-2,5-methano-2H-furo[3,2-b]pyrrolium Bromide (1:1), Scopoline Methobromide (5CI)
SMILES [Br-].C[N+]1(C)[C@@H]2C[C@@H]3C[C@H]1[C@H](O)[C@H]2O3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C9H16NO2.BrH/c1-10(2)6-3-5-4-7(10)9(12-5)8(6)11;/h5-9,11H,3-4H2,1-2H3;1H/q+1;/p-1/t5-,6-,7+,8-,9-;/m0./s1
Formula Weight 249.0364
Chemical Name or Material Scopoline Methobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.