missing translation for 'onlineSavingsMsg'
Learn More

Scopine hydrochloride, MedChemExpress

Product Code. 16408326
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16408326

Brand: Medchem Express HYB0459A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Scopine hydrochloride (6,7-Epoxytropine hydrochloride) is the metabolite of anisodine, which is a α1-adrenergic receptor agonist and used in the treatment of acute circulatory shock.

Scopine hydrochloride (6,7-Epoxytropine hydrochloride) is the metabolite of anisodine, which is a α1-adrenergic receptor agonist and used in the treatment of acute circulatory shock.

Specifications

Chemical Name or Material Scopine hydrochloride
Physical Form Solid
CAS 85700-55-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H14ClNO2
For Use With (Application) Neuroscience-Neuromodulation
Synonym 6,7-Epoxytropine hydrochloride
Solubility Information H2O : ≥ 100 mg/mL (521.76 mM) ∣DMSO : 25 mg/mL (130.44 mM; Need ultrasonic)
SMILES O[C@H]1C[C@H]2[C@@H]3O[C@@H]3[C@H](N2C)C1.[H]Cl
Molecular Weight (g/mol) 191.66
Formula Weight 191.66
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.