missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scopine hydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Scopine hydrochloride (6,7-Epoxytropine hydrochloride) is the metabolite of anisodine, which is a α1-adrenergic receptor agonist and used in the treatment of acute circulatory shock.
Specifications
Specifications
| Chemical Name or Material | Scopine hydrochloride |
| Physical Form | Solid |
| CAS | 85700-55-6 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C8H14ClNO2 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | 6,7-Epoxytropine hydrochloride |
| Solubility Information | H2O : ≥ 100 mg/mL (521.76 mM) ∣DMSO : 25 mg/mL (130.44 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction