missing translation for 'onlineSavingsMsg'
Learn More

SCO2 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18419384 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419384 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419384 Supplier Novus Biologicals Supplier No. NBP177274C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SCO2 Polyclonal antibody specifically detects SCO2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCO2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias cytochrome oxidase deficient homolog 2, MGC125823, MGC125825, protein SCO2 homolog, mitochondrial, SCO cytochrome oxidase deficient homolog 2 (yeast), yeast) homolog 2
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human SCO2. The immunogen is located within the last 50 amino acids of SCO2. Amino Acid Squence: DYYGRSRSAEQISDSVRRH
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9997
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.