missing translation for 'onlineSavingsMsg'
Learn More

SCNN1D Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP1-45221

Product Code. 18257014

  • £329.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SCNN1D Polyclonal specifically detects SCNN1D in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SCNN1D
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
SCNN1D
Peptide with sequence C-AGPQPLETLDT corresponding to C-Terminus according to NP_002969.2.
0.1 mg
Primary
This antibody is specific to the C-Terminus.
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
0.5 mg/mL
Western Blot 1 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 2.5 ug/ml, Peptide ELISA Detection limit 1:16000
Delta-ENaC, Delta-NaCH, DNACH, ENaCd, MGC149710, MGC149711, SCNED, sodium channel, nonvoltage-gated 1, delta, voltage-gated, type I, delta polypeptide
Goat
Affinity Purified
RUO
6339
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only