missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCN9A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534191
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%) Marmoset, Mouse, Panda, Dog, Bovine, Horse, Rabbit, Pig (94%).
Epithelial sodium channels are amiloride-sensitive members of the Degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Members of this superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddels syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1).
Specifications
| SCN9A | |
| Polyclonal | |
| Unconjugated | |
| SCN9A | |
| ETHA; FEB3B; GEFSP7; hNaN; hNE Na; hNE-Na; HSAN2D; KIAA1356; Kiaa4197; MED; mKIAA4197; NAC1; NAC2; NAC3; Nav1.1; Nav1.2; Nav1.3; Nav1.4; Nav1.5; Nav1.6; Nav1.7; Nav1.8; Nav1.9; NE; NE NA; NENA; NE-NA; neuroendocrine sodium channel; Peripheral sodium channel 1; PN1; SCN1; SCN10A; SCN11A; SCN12A; SCN1A; Scn2a; SCN2A1; SCN2A2; SCN3A; SCN4A; SCN5A; SCN8A; Scn9a; SFNP; SNS2; sodium channel 25; sodium channel protein type 9 subunit alpha; Sodium channel protein type IX subunit alpha; sodium channel, voltage gated, type IX alpha subunit; sodium channel, voltage-gated, type 9, alpha polypeptide; sodium channel, voltage-gated, type IX, alpha; sodium channel, voltage-gated, type IX, alpha polypeptide; sodium channel, voltage-gated, type IX, alpha subunit; sodium voltage-gated channel alpha subunit 9; voltage-gated sodium channel alpha subunit Nav1.7; voltage-gated sodium channel subunit alpha Nav1.7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6335 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q15858 | |
| SCN9A | |
| Synthetic 17 amino acid peptide from internal region of human SCN9A / Nav1.7. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction