missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SCN7A Recombinant Protein Antigen

Product Code. 18312807 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 18312807

Brand: Novus Biologicals™ NBP317309PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 6332
Molecular Weight (g/mol) M.W. Theoretical: 27 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias Sodium channel protein cardiac and skeletal muscle subunit alpha, Sodium channel protein type VII subunit alpha, sodium channel, voltage-gated, type VI, alpha, sodium channel, voltage-gated, type VII, alpha, sodium channel, voltage-gated, type VII, alpha polypeptide, voltage-dependent sodium channel alpha subunit, voltage-gated, type VI, alpha polypeptide
Gene Symbol SCN7A
Research Category Neuroscience
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein SCN7A
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.