missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCN7A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550138
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SCN7A protein.
Mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, the protein forms a sodium-selective channel through which Na+ ions may pass in accordance with their electrochemical gradient. George A.L. Jr., Proc. Natl. Acad. Sci. U.S.A. 89:4893-4897(1992).
Specifications
| SCN7A | |
| Polyclonal | |
| Unconjugated | |
| SCN7A | |
| 1110034K09Rik; AI642000; LOW QUALITY PROTEIN: sodium channel protein type 7 subunit alpha; NaG; Na-G; Nav2; Nav2.1; Nav2.2; Nav2.3; Nax; putative voltage-gated sodium channel subunit alpha Nax; Scn6a; SCN7A; Sodium channel protein cardiac and skeletal muscle subunit alpha; sodium channel protein type 7 subunit alpha; Sodium channel protein type VII subunit alpha; sodium channel voltage-gated type VI alpha polypeptide; sodium channel voltage-gated type VII alpha; sodium channel, voltage gated, type VII alpha subunit; sodium channel, voltage-gated, type 6, alpha polypeptide; sodium channel, voltage-gated, type VI, alpha polypeptide; sodium channel, voltage-gated, type VII, alpha; sodium channel, voltage-gated, type VII, alpha subunit; sodium voltage-gated channel alpha subunit 7; voltage-dependent sodium channel alpha subunit | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6332 | |
| -20°C | |
| Liquid |
| Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q01118 | |
| SCN7A | |
| Synthesized peptide derived from internal of human SCN7A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction