Learn More
SCN3B Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25907-1-AP-20UL
This item is not returnable.
View return policy
Description
The SCN3B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SCN3B. This antibody recognizes human, mouse, rat antigen. The SCN3B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| SCN3B | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| HSA243396, KIAA1158, SCN3B, SCNB3, Sodium channel subunit beta 3 | |
| 20 μL | |
| 55800 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |