missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCN3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534188
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Marmoset (100%) Gibbon, Monkey (95%) Dog (89%) Panda, Bovine, Bat, Rabbit (84%).
Mediates the voltage-dependent sodium ion permeability of excitable membranes. Assuming opened or closed conformations in response to the voltage difference across the membrane, the protein forms a sodium-selective channel through which Na+ ions may pass in accordance with their electrochemical gradient.
Specifications
| SCN3A | |
| Polyclonal | |
| PBS with 0.1% sodium azide | |
| Q9NY46 | |
| Scn3a | |
| Synthetic 19 amino acid peptide from C-terminus of human SCN3A / Nav1.3. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Scn3a | |
| BIII; brain III voltage-gated sodium channel; Gm1000; KIAA1356; NAC3; Nav1.3; SCIII; Scn2a; Scn3a; Sodium channel protein brain III subunit alpha; sodium channel protein type 3 subunit alpha; Sodium channel protein type III subunit alpha; sodium channel protein, brain III subunit alpha; sodium channel, voltage gated, type III alpha subunit; sodium channel, voltage-gated, type 3, alpha subunit; sodium channel, voltage-gated, type III, alpha; sodium channel, voltage-gated, type III, alpha polypeptide; sodium channel, voltage-gated, type III, alpha subunit; sodium voltage-gated channel alpha subunit 3; voltage-gated sodium channel subtype III; Voltage-gated sodium channel subunit alpha Nav1.3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6328 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction