missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCN3A, Mouse anti-Human, Clone: 3F3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006328-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: LCESGEMDALRIQMEDRFMASNPSKVSYEPITTTLKRKQEEVSAAIIQRNFRCYLLKQRLKNISSNYNKEAIKGRIDLPIKQDMIIDKLNGNSTPEKTDGSpecifications
| SCN3A | |
| Monoclonal | |
| Unconjugated | |
| sodium channel, voltage-gated, type III, alpha subunit | |
| KIAA1356/NAC3/Nav1.3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3F3 | |
| In 1x PBS, pH 7.4 | |
| NM_006922 | |
| SCN3A | |
| SCN3A (NP_008853, 1861 a.a. ∼ 1960 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6328 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction