missing translation for 'onlineSavingsMsg'
Learn More

SCN3A Antibody, Novus Biologicals™

Product Code. 18332362
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332362 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18332362 Supplier Bio-Techne Supplier No. NBP314469

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SCN3A Polyclonal antibody specifically detects SCN3A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCN3A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias brain III voltage-gated sodium channel, KIAA1356, NAC3, Nav1.3, Sodium channel protein brain III subunit alpha, sodium channel protein type 3 subunit alpha, Sodium channel protein type III subunit alpha, sodium channel, voltage-gated, type III, alpha polypeptide, sodium channel, voltage-gated, type III, alpha subunit, Voltage-gated sodium channel subtype III, Voltage-gated sodium channel subunit alpha Nav1.3
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from internal region of human SCN3A.
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6328
Target Species Human
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Product Type Antibody
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.