missing translation for 'onlineSavingsMsg'
Learn More

SCN3A Antibody (3F3), Novus Biologicals™

Product Code. 18366109 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366109 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18366109 Supplier Novus Biologicals Supplier No. H00006328M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SCN3A Monoclonal antibody specifically detects SCN3A in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCN3A
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3F3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_008853
Gene Alias brain III voltage-gated sodium channel, KIAA1356, NAC3, Nav1.3, Sodium channel protein brain III subunit alpha, sodium channel protein type 3 subunit alpha, Sodium channel protein type III subunit alpha, sodium channel, voltage-gated, type III, alpha polypeptide, sodium channel, voltage-gated, type III, alpha subunit, Voltage-gated sodium channel subtype III, Voltage-gated sodium channel subunit alpha Nav1.3
Host Species Mouse
Immunogen SCN3A (NP_008853, 1861 a.a. ~ 1960 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LCESGEMDALRIQMEDRFMASNPSKVSYEPITTTLKRKQEEVSAAIIQRNFRCYLLKQRLKNISSNYNKEAIKGRIDLPIKQDMIIDKLNGNSTPEKTDG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6328
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.