Learn More
SCN1B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11484-1-AP-150UL
Description
The SCN1B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SCN1B. This antibody recognizes human, mouse, rat antigen. The SCN1B antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| SCN1B | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 6324 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 267 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |