missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCN11A Antibody (Wuhan U. patent anti-Nav1.9) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28782JF585
This item is not returnable.
View return policy
Description
SCN11A Monoclonal antibody specifically detects SCN11A in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| SCN11A | |
| Monoclonal | |
| Janelia Fluor 585 | |
| hNaN, NaN, Nav1.9, Peripheral nerve sodium channel 5, PN5, SCN12A, SNS2, SNS-2, sodium channel, voltage-gated, type XI, alpha polypeptide, sodium channel, voltage-gated, type XI, alpha subunit, sodium channel, voltage-gated, type XII, alpha, voltage-gated sodium channel Nav1.9, Voltage-gated sodium channel subunit alpha Nav1.9, voltage-gated, type XII, alpha polypeptide | |
| SCN11a/ Nav1.9 | |
| 0.1 mL | |
| Signal Transduction | |
| 11280 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Wuhan U. patent anti-Nav1.9 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction