missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SCN11A Antibody (Wuhan U. patent anti-Nav1.9) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28782-100ug
This item is not returnable.
View return policy
Description
SCN11A Monoclonal antibody specifically detects SCN11A in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| SCN11A | |
| Monoclonal | |
| Unconjugated | |
| Q9UI33 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Wuhan U. patent anti-Nav1.9 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| hNaN, Nav1.9, Peripheral nerve sodium channel 5, PN5, SCN12A, SNS2, SNS-2, sodium channel, voltage-gated, type XI, alpha polypeptide, sodium channel, voltage-gated, type XI, alpha subunit, sodium channel, voltage-gated, type XII, alpha, voltage-gated sodium channel Nav1.9, Voltage-gated sodium channel subunit alpha Nav1.9, voltage-gated, type XII, alpha polypeptide | |
| SCN11a/ Nav1.9 | |
| 100 μg | |
| Signal Transduction | |
| 11280 | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido