missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SCN11A Monoclonal antibody specifically detects SCN11A in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | SCN11A |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 6E1 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_054858 |
| Gene Alias | hNaN, NaN, Nav1.9, Peripheral nerve sodium channel 5, PN5, SCN12A, SNS2, SNS-2, sodium channel, voltage-gated, type XI, alpha polypeptide, sodium channel, voltage-gated, type XI, alpha subunit, sodium channel, voltage-gated, type XII, alpha, voltage-gated sodium channel Nav1.9, Voltage-gated sodium channel subunit alpha Nav1.9, voltage-gated, type XII, alpha polypeptide |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?