missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCML2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584666
This item is not returnable.
View return policy
Description
Immunogen sequence: SRSVPGTTSS PLVGDISPKS SPHEVKFQMQ RKSEAPSYIA VPDPSVLKQG FSKDPSTWSV DEVIQFMKHT DPQISGPLAD LFR Highest antigen sequence indentity to the following orthologs: Rat - 58%, Mouse - 57%.
SCML2 is a putative component of a Polycomb group (PcG) multiprotein complex involved in represing transcription of homeotic genes during development. It mediates chromatin remodeling and DNA dependent negative regulation of transcription by binding to methylated lysine residues in histone tails.
Specifications
| SCML2 | |
| Polyclonal | |
| Unconjugated | |
| SCML2 | |
| 4932420G07Rik; Scm polycomb group protein like 2; SCML2; sex comb on midleg-like 2 (Drosophila); sex comb on midleg-like protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10389 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UQR0 | |
| SCML2 | |
| Recombinant protein corresponding to Human SCML2. Recombinant protein control fragment (Product #RP-107363). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction