missing translation for 'onlineSavingsMsg'
Learn More

SCML2 Antibody (OTI1E1), DyLight 594, Novus Biologicals™

Product Code. 18415996 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415996 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415996 Supplier Novus Biologicals Supplier No. NBP274024DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SCML2 Monoclonal antibody specifically detects SCML2 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCML2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1E1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias sex comb on midleg (Drosophila)-like 2, sex comb on midleg-like 2 (Drosophila), sex comb on midleg-like protein 2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-321 of human SCML2 (NP_006080) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10389
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.