missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCLY Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563172
This item is not returnable.
View return policy
Description
Immunogen sequence: LYPMLFGGGQ ERNFRPGTEN TPMIAGLGKA AELVTQNCEA YEAHMRDVRD YLEERLEAEF GQKRIHLNS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.
The SCLY gene encodes selenocysteine lyase, an enzyme involved in selenium metabolism, which plays a critical role in the recycling of selenocysteine from selenoproteins. SCLY catalyzes the conversion of selenocysteine into alanine, releasing selenium for reuse in the biosynthesis of new selenoproteins. It is chiefly active in the liver and kidneys, contributing to maintaining selenium homeostasis, which is vital for numerous biological functions due to selenium's role in antioxidant defense and thyroid hormone metabolism. Alterations in SCLY function or expression can affect selenium levels in the body and are associated with disorders related to oxidative stress and metabolic imbalances. Given the importance of selenium in cellular protection and immune function, SCLY is also of interest in studies focusing on dietary supplementation and disease prevention strategies, particularly in conditions like cancer and cardiovascular diseases where oxidative damage is prevalent.
Specifications
| SCLY | |
| Polyclonal | |
| Unconjugated | |
| Scly | |
| 9830169H08; A930015N15Rik; hSCL; mSCL; SCL; SCLY; Scly1; Scly2; Selenocysteine lyase; selenocysteine lyase; Selenocysteine reductase; selenocysteine reductase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51540 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96I15 | |
| Scly | |
| Recombinant protein corresponding to Human SCLY. Recombinant protein control fragment (Product #RP-103918). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction