missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SCLY Polyclonal antibody specifically detects SCLY in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
Specifications
| Antigen | SCLY |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000 - 1:10000, Immunoprecipitation 20 ug/mg lysate |
| Formulation | Tris-citrate/phosphate pH 7 -8. |
| Gene Alias | EC 4.4.1.16, SCLhSCL, selenocysteine lyase |
| Host Species | Rabbit |
| Immunogen | Recognizes region between residues Between Between 395 and 445.. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?