missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCL/Tal1 Antibody (OTI5H1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74022CL1
This item is not returnable.
View return policy
Description
SCL/Tal1 Monoclonal antibody specifically detects SCL/Tal1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| SCL/Tal1 | |
| Monoclonal | |
| CoraFluor 1 | |
| BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product | |
| Full length human recombinant protein of human TAL1(NP_003180) produced in HEK293T cell. | |
| 0.1 mL | |
| Phospho Specific | |
| 6886 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI5H1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction