missing translation for 'onlineSavingsMsg'
Learn More

SCL/Tal1 Antibody (333803) [HRP], Novus Biologicals™

Product Code. 30736573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736573 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736573 Supplier Novus Biologicals Supplier No. FAB3360H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SCL/Tal1 Monoclonal antibody specifically detects SCL/Tal1 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SCL/Tal1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 333803
Conjugate HRP
Formulation PBS
Gene Alias BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product
Host Species Mouse
Immunogen E. coli-derived recombinant human SCL/Tal1, Arg199-Gly266, Accession # P17542
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6886
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.