missing translation for 'onlineSavingsMsg'
Learn More

SCL/Tal1 Antibody (1E6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£150.00 - £307.00

Specifications

Antigen SCL/Tal1
Clone 1E6
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Applications Western Blot, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18608606
View Documents
Novus Biologicals
NBP2-50285-0.025ml
0.025 mL
£150.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18624777
View Documents
Novus Biologicals
NBP2-50285
0.1 mL
£307.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SCL/Tal1 Monoclonal antibody specifically detects SCL/Tal1 in Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

SCL/Tal1
1 mg/mL
Western Blot, Immunofluorescence
Unconjugated
Mouse
Phospho Specific
PBS
6886
IgG1
Protein A purified
1E6
Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Monoclonal
Purified
RUO
Zebrafish
BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product
GST-SCL/Tal1 (N-terminal) fusion protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.