missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SCIO 469 hydrochloride
Description
SCIO 469 hydrochloride is a selective, ATP-competitive p38 inhibitor (IC50 = 9 nM for p38α in vitro). Displays approximately 10-fold selectivity for p38α over p38β and 2000-fold selectivity for p38α over 20 other kinases. Reduces p38α phosphorylation in multiple myeloma cells in vitro and in vivo; activity results in decreased tumor burden and angiogenesis in murine models of multiple myeloma. Also enhances bortezomib -induced cytotoxicity against multiple myeloma cells.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 549.46 |
| Purity | >98% |
| Molecular Weight (g/mol) | 513.01 |
| Chemical Name or Material | SCIO 469 hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?