missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Scintillation Vials

Product Code. 31923410
Change view
Click to view available options
Volume (Metric):
4 mL
6.5 mL
20 mL
Height (Metric):
53 mm
57 mm
61.4 mm
Diameter (Metric) Outer:
13.8 mm
16 mm
26.5 mm
Unit Size:
Pack of 1000
Pack of 2000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Height (Metric) Diameter (Metric) Outer Unit Size
31923410 20 mL 61.4 mm 26.5 mm Pack of 1000
31922506 4 mL 53 mm 13.8 mm Pack of 2000
31922796 6.5 mL 57 mm 16 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31923410 Supplier Globe Scientific Supplier No. 101010
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to provide consistently low background counts and are suitable for low activity counting.

20mL Scintillation Vial:

  • Scintillation counting, gamma counting, chromatography, sample storage and culturing
  • Because of its leak-tight cap it is also commonly used to store and transport histological specimens in 10% formalin
  • Provides high degree of resistance to organic solvents and sufficient translucence so that reagent levels can be easily seen
  • Unique thread design of closure permits vial to be sealed tight with easy 1/4 turn and with minimum pressure

6.5mL Scintillation Vial:

  • Securely sealed by simply snapping caps on
  • Cap is conveniently removed with easy 1/4 turn to left

4mL Polyethylene Scintillation Vial:

  • Uses full turn polyethylene screw cap
  • Most economical size to use because it requires least amount of solvent

Specifications

Diameter (Metric) Outer 26.5 mm
Material HDPE
Sterility Non-sterile
Volume (Metric) 20 mL
No. per Case 1000
Closure Material Polyethylene
Closure Type Screw Cap
Height (Metric) 61.4 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.