missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCIN Antibody (12B11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26334-100ul
This item is not returnable.
View return policy
Description
SCIN Monoclonal antibody specifically detects SCIN in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| SCIN | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 12B11 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| KIAA1905adseverin, Scinderin | |
| A synthesized peptide derived from Human SCIN [UniProt Q9Y6U3] | |
| 100 μL | |
| Apoptosis | |
| 85477 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction