Learn More
Cole-Parmer™ Scientific Industries™ Vortex-Genie™ Vortex Shaker Platform Head with Cover
Product Code. 11749669 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (Metric):
7.62 cm
Unit Size:
Each
Product Code. Diameter (Metric) Unit Size
11749669 7.62 cm Each
1 options
Product Code. 11749669 Supplier Cole-Parmer™ Supplier No. 0472461
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Maximize high-speed touch mixing
  • Vortex Shaker Platform Head
  • with Cover
  • 3 inch Diameter
  • For use with Scientific Industries™ Vortex-Genie™
Specifications
Product Type Vortex Shaker Platform Head with Cover
Diameter (Metric) 7.62 cm
For Use With (Equipment) Scientific Industries™ Vortex-Genie™
Includes Cover