Learn More
Cole-Parmer™ Scienceware™ Stainless Steel Vibrating Spatula with PTFE Coating
Shop All Cole-Parmer ProductsDescription
- Move the toothed thumbwheel to vibrate the blade and add as little or as much of your material as needed
- Narrow V-shaped blade maneuvers easily into small-mouth containers and tight corners
- Stainless steel blade is also available with a brown PTFE coating
- FDA compliant for direct food contact applications
Specifications
Specifications
| Product Type | Vibrating Spatula |
| Length (English) | 8.25 in. |
| Length (Metric) Blade | 20.96 cm |
| Material | Stainless Steel/PTFE Coated |