Learn More
SI Analytics™ ScienceLine Ion Selective Electrode
Product Code. 10516063 Shop All SI Analytics Products
Click to view available options
Parameter:
Bromide
Chloride
Copper
Iodide
Potassium
Sulfide
pH Range:
0 to 14
1 to 12
2 to 12
2 to 6
Unit Size:
Each
Product Code. 10516063
Brand: SI Analytics™ 285130410
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
SI Analytics ScienceLine Ion Selective Electrodes possess the advantage of a much lower temperature sensitivity and a metal ion free reference system.
  • ScienceLine sensors for ammonia, sodium, oxygen, ion-selective indicator electrodes
  • Also measure pH and temperature
TRUSTED_SUSTAINABILITY
Specifications
pH Range 0 to 14
Model I 60
Electrode Selective
Temperature (Metric) Range 0°C to 80°C
Type Ion Selective Electrode
Measurement Range 0.006 to 127,000 mg/L
Parameter Iodide