missing translation for 'onlineSavingsMsg'
Learn More

Porvair Sciences™ Science Polystyrene High Bind White Microplate

Product Code. 12364108
Encompass_Preferred
Change view
Click to view available options
No. of Wells:
384
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells unitSize
12364108 384 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12364108 Supplier Porvair Sciences™ Supplier No. 222012

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Square wells maximise liquid volume per well (120μL/well)
  • White plate prevents cross talk in light emitting assays
  • Black plate gives low autofluorescence background
  • Tissue culture treated plates for anchorage dependent cells
  • Total well volume 120μL
  • Dimensions [w x d x h] 3.75mm x 3.75mm x 11.4mm (well), 85.5mm x 127.5mm x 14.8mm (plate)
  • 24 x 16 well array

Specifications

Format Standard
Array 16 x 24
Color White
Material Polystyrene
No. of Wells 384
Total Volume (Metric) Well 120 μL
No. per Pack 100
Quantity 100 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.