missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ School Grade Filter Paper

Provides for general filtration in the school environment. Macherey Nagel™ School Grade Filter Paper is manufactured from cellulose.

Brand:  Macherey-Nagel™ 4007130180

10.43 GBP valid until 2024-04-30
Use promo code "21637" to get your promotional price.



Code : 02

Additional Details : Weight : 0.19620kg

 View more versions of this product

Product Code. 12982701

  • £30.10 / Pack of 100
In Stock
to see stock.
Add to basket

Description

Description

  • General purpose
  • Available in a range of sizes
Specifications

Specifications

Cellulose
School grade
100 Pack
180 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers