missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Schisandrol B, MedChemExpress
Description
Schisandrol B (Gomisin-A) is a major active constituent of Schisandra sphenanthera with hepato-protective effects. Schisandrol B inhibits reactive oxygen species (ROS) production. Schisandrol B inhibits the activity of P-glycoprotein and CYP3A and also has anti-inflammatory, anti-diabetic and antioxidant activities.
Specifications
Specifications
| Chemical Name or Material | Schisandrol B |
| CAS | 58546-54-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H28O7 |
| Quantity | 10 mg |
| Synonym | Gomisin-A TJN-101 Wuweizi alcohol-B |
| Solubility Information | DMSO : 50 mg/mL (120.06 mM; Need ultrasonic) |
| SMILES | O[C@@]([C@@H](C)C1)(C)CC2=CC(OC)=C(OC)C(OC)=C2C3=C1C=C4OCOC4=C3OC |
| Molecular Weight (g/mol) | 416.46 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?