missing translation for 'onlineSavingsMsg'
Learn More

Schiff's Reagent

Product Code. 17897446
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging
17897446 500 mL Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17897446 Supplier Thermo Scientific Alfa Aesar Supplier No. J62171.AP
  • ONLINE EXCLUSIVE PRICE
    £124.44
    £100.65 / 500mL
    Save £23.79
    19% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 569-61-9 | C19H18ClN3 | 323.824 g/mol

Intended for use in Sigma Periodic Acid Staining Procedure, 395. Kit is intended for use with blood, bone marrow, tissue touch preparations or routine tissue sections. It is used for staining glycoproteins in polyacrylamide gels. Schiff's reagent is also sometimes used to dye organic tissues. It is been widely used for detection of a certain functional group in chemical solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 569-61-9
Packaging Plastic bottle
Molecular Formula C19H18ClN3
MDL Number MFCD00081985
Quantity 500 mL
Sensitivity Air and light sensitive
Synonym basic red 9, pararosaniline hydrochloride, parafuchsin, pararosaniline chloride, parafuchsine, p-fuchsin, basic fuchsin, ci basic red 9, c.i. basic red 9, parafuksin
Solubility Information It is soluble in water and methanol.
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
SMILES C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl
IUPAC Name 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
Molecular Weight (g/mol) 323.824
PubChem CID 11292
ChEBI CHEBI:87663
Physical Form Liquid
Chemical Name or Material Schiff's Reagent
Show More Show Less
compliance-icons
Product Identifier
  • Schiff's Reagent
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium metabisulfite, Magenta, Hydrochloric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.