missing translation for 'onlineSavingsMsg'
Learn More

SCH 58261, MedChemExpress

Product Code. 16407636
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16407636

Brand: Medchem Express HY19533

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SCH 58261 is a potent, selective and competitive antagonist of adenosine A2A receptor with an IC50 of 15 nM, and displays 323-, 53- and 100-fold more selective for A2A receptor than A1, A2B, and A3 receptors, respectively.

SCH 58261 is a potent, selective and competitive antagonist of adenosine A2A receptor with an IC50 of 15 nM, and displays 323-, 53- and 100-fold more selective for A2A receptor than A1, A2B, and A3 receptors, respectively.

Specifications

Chemical Name or Material SCH 58261
Physical Form Solid
CAS 160098-96-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H15N7O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 34 mg/mL (98.45 mM)
SMILES NC1=NC(N(CCC2=CC=CC=C2)N=C3)=C3C4=NC(C5=CC=CO5)=NN14
Molecular Weight (g/mol) 345.36
Formula Weight 345.36
Percent Purity 99.62%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.