missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SCH 23390 hydrochloride
Description
Potent dopamine receptor antagonist (Ki values are 0.2 nM and 0.3 nM at D1 and D5 receptor sub-types, respectively). Also an agonist at 5-HT2C receptors in vitro (Ki values are 6.3 - 9.3 nM). Blocks quinpirole-induced Kir3 (GIRK) currents (EC50 = 268 nM) independently of receptors.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 324.24 |
| Purity | >98% |
| Molecular Weight (g/mol) | 324.245 |
| Chemical Name or Material | SCH 23390 hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?