missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SCH 23390 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent dopamine receptor antagonist (Ki values are 0.2 nM and 0.3 nM at D1 and D5 receptor sub-types, respectively). Also an agonist at 5-HT2C receptors in vitro (Ki values are 6.3 - 9.3 nM). Blocks quinpirole-induced Kir3 (GIRK) currents (EC50 = 268 nM) independently of receptors.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 324.24 |
| Purity | >98% |
| Molecular Weight (g/mol) | 324.245 |
| Chemical Name or Material | SCH 23390 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction