missing translation for 'onlineSavingsMsg'
Learn More

SCH-23390 hydrochloride, MedChemExpress

Product Code. 16440268
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16440268

Brand: Medchem Express HY19545A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SCH-23390 hydrochloride (R-(+)-SCH-23390 hydrochloride) is a potent and selective dopamine D1-like receptor antagonist with Kis of 0.2 nM and 0.3 nM for the D1 and D5 receptor, respectively.

SCH-23390 hydrochloride is a potent and high efficacy human 5-HT2C receptor agonist with a Ki of 9.3 nM. SCH-23390 hydrochloride also binds with high affinity to the 5-HT2 and 5-HT1C receptors. SCH-23390 hydrochloride inhibits G protein-coupled inwardly rectifying potassium (GIRK) channels with an IC50 of 268 nM.

Specifications

Chemical Name or Material SCH-23390 hydrochloride
CAS 125941-87-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H19Cl2NO
Quantity 5 mg
Synonym R-(+)-SCH-23390 hydrochloride
Solubility Information DMSO : ≥ 32 mg/mL (98.69 mM) ∣H2O : 28.57 mg/mL (88.11 mM; Need ultrasonic)
SMILES OC1=C(Cl)C=C2CCN(C)C[C@H](C3=CC=CC=C3)C2=C1.[H]Cl
Molecular Weight (g/mol) 324.24
Formula Weight 324.24
Percent Purity 99.21%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.