missing translation for 'onlineSavingsMsg'
Learn More

SCH-1473759, MedChemExpress

Product Code. 16494996
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494996 100 mg 100mg
16476707 5 mg 5mg
16453998 10 mg 10mg
16430116 2 mg 2mg
16400357 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494996 Supplier Medchem Express Supplier No. HY10482
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SCH-1473759 is an aurora inhibitor with IC50s of 4 and 13 nM for aurora A and B, respectively.

SCH-1473759 is an aurora inhibitor with IC50s of 4 and 13 nM for aurora A and B, respectively.

Specifications

Chemical Name or Material SCH-1473759
Physical Form Solid
CAS 1094069-99-4
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H26N8OS
For Use With (Application) Cancer-Kinase/protease
SMILES CC1=CN2C(C(NC3=CC(CN(C(C)(CO)C)CC)=NS3)=N1)=NC=C2C4=CNN=C4
Molecular Weight (g/mol) 426.54
Formula Weight 426.54
Percent Purity 98.2%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.