missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCGN Polyclonal Antibody
Chicken Polyclonal Antibody
Brand: Invitrogen™ PA5143546
This item is not returnable.
View return policy
Description
SCGN Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IHC (PFA)
The encoded protein is a secreted calcium-binding protein which is found in the cytoplasm. It is related to calbindin D-28K and calretinin. This protein is thought to be involved in KCL-stimulated calcium flux and cell proliferation.
Specifications
| SCGN | |
| Polyclonal | |
| Unconjugated | |
| SCGN | |
| CALBL; DJ501N12.8; SCGN; SECRET; Secretagogin; secretagogin, EF-hand calcium binding protein; SEGN; setagin | |
| Chicken | |
| Affinity chromatography | |
| RUO | |
| 10590, 214189, 306942 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5 mM sodium azide | |
| O76038, Q6R556, Q91WD9 | |
| SCGN | |
| Full-length recombinant human secretagogin protein. | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction