missing translation for 'onlineSavingsMsg'
Learn More

SCGF Mouse anti-Human, Clone: 4H11E4, Proteintech

Product Code. 16827375 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16827375 20 μL 20µL
16817375 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16827375 Supplier Proteintech Supplier No. 602951IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This gene encodes a member of the C-type lectin superfamily. The encoded protein is a secreted sulfated glycoprotein and functions as a growth factor for primitive hematopoietic progenitor cells. An alternative splice variant has been described but its biological nature has not been determined.

Specifications

Antigen SCGF
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4H11E4
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CLEC11A
Gene Accession No. Q9Y240
Gene Alias CLEC11A, CLECSF3, LSLCL, P47, SCGF, Stem cell growth factor
Gene Symbols CLEC11A
Host Species Mouse
Immunogen SCGF Fusion Protein Ag21365
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6320
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.