missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCGF Mouse anti-Human, Clone: 4H11E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60295-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the C-type lectin superfamily. The encoded protein is a secreted sulfated glycoprotein and functions as a growth factor for primitive hematopoietic progenitor cells. An alternative splice variant has been described but its biological nature has not been determined.Specifications
| SCGF | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y240 | |
| CLEC11A | |
| SCGF Fusion Protein Ag21365 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 4H11E4 | |
| Unconjugated | |
| CLEC11A | |
| CLEC11A, CLECSF3, LSLCL, P47, SCGF, Stem cell growth factor | |
| Mouse | |
| Protein A | |
| RUO | |
| 6320 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction