missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCGF/CLEC11a Antibody (MM0533-6D5), mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
SCGF/CLEC11a Monoclonal antibody specifically detects SCGF/CLEC11a in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Formulation | 50mM Sodium Borate |
| Quantity | 0.1 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?