missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SCGB1D2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550697
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SCGB1D2 protein.
May bind androgens and other steroids, may also bind estramustine, a chemotherapeutic agent used for prostate cancer. May be under transcriptional regulation of steroid hormones.
Specifications
| SCGB1D2 | |
| Polyclonal | |
| Unconjugated | |
| SCGB1D2 | |
| LIPB; LIPHB; Lipophilin-B; LPHB; LPNB; prostatein-like lipophilin B; SCGB1D2; Secretoglobin family 1D member 2; secretoglobin, family 1D member 2; secretoglobin, family 1D, member 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10647 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 2.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O95969 | |
| SCGB1D2 | |
| Synthetic peptide corresponding to residues near the C terminal of human secretoglobin, family 1D, member 2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction