Learn More
Invitrogen™ SCG3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121140
Description
Positive test controls include: 22Rv1, HepG2, Mouse brain, Mouse lung, Mouse heart, Rat brain, Rat lung, Rat heart. The target is usually found in the following locations: Cytoplasmic vesicle, Peripheral membrane protein, Secreted, secretory vesicle lumen, secretory vesicle membrane. Immunogen sequence: KLISKEANNY EEDPNKPTSW TENQAGKIPE KVTPMAAIQD GLAKGENDET VSNTLTLTNG LERRTKTYSE DNFEELQYFP NFYALLKSID SEKEAKEKET LITIMKTLID FVKMMVKYGT ISPEEGVSYL ENLDEMIALQ TKNKLEKNAT DNISKLFPAP SEKSHEETDS TKEEAAKMEK EYGSLKDSTK DDNSNPGGKT DEPKGKTEAY LEAIRKNIEW LKKHDKKGNK EDYDLSKMRD FINKQADAYV EKGILDKEEA EAIKRIYSSL.
The protein encoded by this gene is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. Granins may serve as precursors for biologically active peptides. Some granins have been shown to function as helper proteins in sorting and proteolytic processing of prohormones; however, the function of this protein is unknown. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| SCG3 | |
| Polyclonal | |
| Unconjugated | |
| SCG3 | |
| 1B1075; AI385542; Chgd; Scg3; Secretogranin III; secretogranin-3; SgIII; UNQ2502/PRO5990 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116635, 20255, 29106 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.09 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P47867, P47868, Q8WXD2 | |
| SCG3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 199-468 of human SCG3 (NP_037375.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.