missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCFD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13375-1-AP-20UL
This item is not returnable.
View return policy
Description
SCFD2 may be involved in protein transport.Specifications
| SCFD2 | |
| Polyclonal | |
| Unconjugated | |
| SCFD2 | |
| SCFD2, STXBP1L1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 152579, 212986, 498353 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BTY8, Q8WU76 | |
| Scfd2 | |
| SCFD2 Fusion Protein Ag4197 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction